Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000557090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
ENST00000557090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000557090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
ENST00000557090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010650.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
ENST00000435624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
ENST00000495064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
NR_104113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400180561.hg.1 |
TC1400010650.hg.1 |
Clariom D TC |