Assay Details
Gene Symbol: LOC100996447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104147.1 | 1 - 2 | 896 | 120 | ||
GenBank mRNA | AK097794.1 | - | 1 - 2 | 896 | 120 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | arginine and serine rich coiled-coil 1 | RSRC1 |
BM-011, SFRS21, SRrp53 |
-23.1kb | Anti-sense | Chr. 3 - 158110052 - 158544835 on NCBI Build GRCh38 |
Downstream | myeloid leukemia factor 1 | MLF1 | 97.0 | Anti-sense | Chr. 3 - 158571162 - 158606460 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196700.hg.1 | TC0300012939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196702.hg.1 | TC0300012939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196706.hg.1 | TC0300012939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196705.hg.1 | TC0300012939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196704.hg.1 | TC0300012939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | ENST00000479233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012939.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | ENST00000475981.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | NR_104147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300196703.hg.1 | TC0300012939.hg.1 | Clariom D TC |
