Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
ENST00000479233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012939.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196695.hg.1 |
TC0300012939.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
ENST00000475981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
NR_104147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196696.hg.1 |
TC0300012939.hg.1 |
Clariom D TC |