Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000629295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_047469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_047469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_047469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157167.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157167.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157167.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157167.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157167.hg.1 |
TC0300008000.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000628435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000629295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
ENST00000630120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_047469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_047469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_047469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008000.hg.1 |
NR_104153.1 |
Refseq |