Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194733.hg.1 |
TC1100012332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194735.hg.1 |
TC1100012332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194734.hg.1 |
TC1100012332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
ENST00000525984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012332.hg.1 |
NR_104155.1 |
Refseq |