Assay Details
Gene Symbol: LOC283352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104172.1 | 2 - 3 | 218 | 79 | ||
GenBank mRNA | BC035922.1 | - | 1 - 2 | 59 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glycosyltransferase 1 domain containing 1 | GLT1D1 | -126.0kb | Sense | Chr. 12 - 128853323 - 128984964 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101927735 | LOC101927735 | 97.1kb | Sense | Chr. 12 - 129208601 - 129212662 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000510001.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000541320.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | ENST00000542535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009378.hg.1 | NR_104173.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000508925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000512133.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000537367.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | ENST00000540739.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | NR_104172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200169845.hg.1 | TC1200009378.hg.1 | Clariom D TC |
