Assay Details
Gene Symbol: NFIA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104180.1 | 2 - 3 | 186 | 91 | ||
GenBank mRNA | AW172317.1 | - | 2 - 3 | 91 | ||
HY006887.1 | - | 2 - 3 | 189 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NFIA antisense RNA 2 | NFIA-AS2 | -279.2kb | Sense | Chr. 1 - 60940244 - 60970776 on NCBI Build GRCh38 | |
Downstream | sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase) pseudogene | MGC34796 | 402.9kb | Anti-sense | Chr. 1 - 61654242 - 61656128 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000630993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000630993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | ENST00000630993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000617929.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000630993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000630993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | ENST00000630993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014386.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000442027.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000596404.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000626805.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | ENST00000630199.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | NR_104180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207733.hg.1 | TC0100014386.hg.1 | Clariom D TC |
