Assay Details
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104217.1 | 1 - 2 | 32 | 78 | ||
GenBank mRNA | AK125792.1 | - | 1 - 2 | 32 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myomegalin-like | LOC101060254 | -62.0kb | Anti-sense | Chr. 1 - 145506388 - 145567523 on NCBI Build GRCh38 | |
Downstream | G protein-coupled receptor 89A | GPR89A |
GPHR, GPR89, GPR89B, SH120, UNQ192 |
102.0 | Anti-sense | Chr. 1 - 145607960 - 145670977 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216419.hg.1 | TC0100015585.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100216420.hg.1 | TC0100015585.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000437377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000606775.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | ENST00000619932.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015585.hg.1 | NR_104217.1 | Refseq |
