Assay Details
| Gene Symbol: MT1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104046.1 | 1 - 2 | 106 | 122 | ||
| GenBank mRNA | BC171791.1 | - | 1 - 2 | 90 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | metallothionein 1H | MT1H |
MT-0, MT-1H, MT-IH, MT1 |
-5.0kb | Sense | Chr. 16 - 56669814 - 56671129 on NCBI Build GRCh38 |
| Downstream | metallothionein 1X | MT1X |
MT-1l, MT1 |
4.6kb | Sense | Chr. 16 - 56682470 - 56684196 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

MT1IP
Genomic Map