Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010589.hg.1 |
ENST00000413430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010589.hg.1 |
ENST00000413430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010589.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010589.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010589.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177204.hg.1 |
ENST00000413430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177204.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177204.hg.1 |
ENST00000413430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177204.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177204.hg.1 |
TC0300010589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300177205.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177205.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177205.hg.1 |
TC0300010589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300177206.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177206.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177206.hg.1 |
TC0300010589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010589.hg.1 |
ENST00000413430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010589.hg.1 |
ENST00000413430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010589.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010589.hg.1 |
NR_109804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010589.hg.1 |
NR_109804.1 |
Refseq |