Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009889.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169369.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169369.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169369.hg.1 |
TC2000009889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169369.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169369.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009889.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169368.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169368.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169368.hg.1 |
TC2000009889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169368.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169368.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169370.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169370.hg.1 |
NR_109861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169370.hg.1 |
TC2000009889.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169370.hg.1 |
ENST00000449270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169370.hg.1 |
NR_109861.1 |
Refseq |