Assay Details
| Gene Symbol: ID2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110153.1 | 2 | 714 | 67 | ||
| GenBank mRNA | AA970710.1 | - | 1 | 67 | ||
| AI034463.1 | - | 1 | 67 | |||
| DB336412.1 | - | 1 | 67 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929567 | LOC101929567 | -96.0kb | Sense | Chr. 2 - 8559833 - 8583792 on NCBI Build GRCh38 | |
| Downstream |
inhibitor of DNA binding 2, HLH protein |
ID2 |
GIG8, ID2A, ID2H, bHLHb26 |
1.4kb | Anti-sense | Chr. 2 - 8681983 - 8684453 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

ID2-AS1
Genomic Map