Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009009.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163882.hg.1 |
TC0700009009.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700163880.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163880.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163880.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163880.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163880.hg.1 |
TC0700009009.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000413812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
ENST00000450061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009009.hg.1 |
NR_110195.1 |
Refseq |