Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147435.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100147435.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147435.hg.1 |
TC2100007025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100147435.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100147435.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007025.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147436.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100147436.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147436.hg.1 |
TC2100007025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100147436.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100147436.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147437.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100147437.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147437.hg.1 |
TC2100007025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100147437.hg.1 |
ENST00000457157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100147437.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147438.hg.1 |
NR_110419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100147438.hg.1 |
TC2100007025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100147438.hg.1 |
NR_110419.1 |
Refseq |