Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011136.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011136.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011136.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011136.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011136.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011136.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193226.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193226.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193226.hg.1 |
TC1900011136.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193226.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193226.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193225.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193225.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193225.hg.1 |
TC1900011136.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193225.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193225.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193227.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193227.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193227.hg.1 |
TC1900011136.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193227.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193227.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011136.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011136.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011136.hg.1 |
ENST00000599433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011136.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011136.hg.1 |
NR_110729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011136.hg.1 |
NR_110729.1 |
Refseq |