Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181658.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181658.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181658.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181658.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181658.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100181659.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181659.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181659.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181659.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181659.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100181649.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181649.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181649.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181649.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181649.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100181651.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181651.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181651.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181651.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181651.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100181652.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181652.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181652.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181652.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181652.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100181657.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181657.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181657.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181657.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181657.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100181660.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181660.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181660.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181660.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181660.hg.1 |
TC0100011144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
ENST00000568695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011144.hg.1 |
NR_110787.1 |
Refseq |