Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010381.hg.1 |
ENST00000625644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147265.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147265.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147265.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147265.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147265.hg.1 |
TC1300006969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010381.hg.1 |
ENST00000625644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147252.hg.1 |
TC1300006969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006969.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147256.hg.1 |
TC1300006969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147251.hg.1 |
TC1300006969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000615685.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000616251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000619407.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
ENST00000620300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
NR_120423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147254.hg.1 |
TC1300006969.hg.1 |
Clariom D TC |