| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157943.hg.1 |
TC0800008434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157944.hg.1 |
TC0800008434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157945.hg.1 |
TC0800008434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000499653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008434.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157941.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157941.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157941.hg.1 |
ENST00000520820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157941.hg.1 |
NR_125415.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157941.hg.1 |
TC0800008434.hg.1 |
Clariom D TC |