Assay Details
| Gene Symbol: LINC01565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125802.1 | 3 | 1626 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC90246 | LOC90246 | -61.4kb | Anti-sense | Chr. 3 - 128507835 - 128510595 on NCBI Build GRCh38 | |
| Downstream | ribophorin I | RPN1 |
OST1, RBPH1 |
45.7kb | Sense | Chr. 3 - 128619970 - 128650876 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01565
Genomic Map