Assay Details
Gene Symbol: LOC102724804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125828.1 | 1 | 219 | 117 | ||
GenBank mRNA | DB073909.1 | - | 1 | 219 | 117 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | phosphatidylserine synthase 1 | PTDSS1 |
LMHD, PSS1, PSSA |
-52.6kb | Anti-sense | Chr. 8 - 96261886 - 96334552 on NCBI Build GRCh38 |
Downstream | syndecan 2 | SDC2 |
CD362, HSPG, HSPG1, SYND2 |
105.9kb | Anti-sense | Chr. 8 - 96493654 - 96611809 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176144.hg.1 | TC0800011177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800176149.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176149.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176149.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176149.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176149.hg.1 | TC0800011177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176146.hg.1 | TC0800011177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176145.hg.1 | TC0800011177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176143.hg.1 | TC0800011177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000522767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | ENST00000523862.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011177.hg.1 | NR_125829.1 | Refseq |
