Assay Details
| Gene Symbol: COL5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000093.4 | NP_000084.3 | 2 | 597 | 134 | |
| NM_001278074.1 | NP_001265003.1 | 2 | 597 | 134 | ||
| XM_017014266.1 | XP_016869755.1 | 2 | 612 | 134 | ||
| GenBank mRNA | AB209864.1 | - | 1 | 76 | 134 | |
| AB371583.1 | - | 2 | 597 | 134 | ||
| D90279.1 | - | 2 | 340 | 134 | ||
More Information
Set Membership: |
|

COL5A1
Genomic Map