Assay Details
| Gene Symbol: PRDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182907.2 | NP_878911.1 | 1 | 123 | 65 | |
| GenBank mRNA | AK301341.1 | - | 1 | 123 | 65 | |
| AY198415.1 | - | 1 | 45 | 65 | ||
| BC103832.1 | - | 1 | 36 | 40001920 | 65 | |
| BC103833.1 | - | 1 | 36 | 40001921 | 65 | |
| BC103834.1 | - | 1 | 36 | 40001922 | 65 | |
| BC103835.1 | - | 1 | 36 | 40001923 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000844.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK301341.1,AY198415.1,BC103832.1,BC103833.1,BC103834.1,BC103835.1,NM_182907.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
morphogenesis of a branching structure
maternal placenta development
heart valve development
ventricular septum development
transcription, DNA-templated
germ cell development
post-embryonic development
positive regulation of gene expression
negative regulation of B cell proliferation
negative regulation of lipopolysaccharide-mediated signaling pathway
methylation
regulation of natural killer cell differentiation
regulation of extrathymic T cell differentiation
aorta development
eye photoreceptor cell development
cell fate commitment
positive regulation of B cell differentiation
artery morphogenesis
regulation of NK T cell differentiation
intestinal epithelial cell development
trophoblast giant cell differentiation
coronary vasculature development
sebum secreting cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
methyltransferase activity
histone deacetylase binding
metal ion binding
promoter-specific chromatin binding
negative regulation of transcription from RNA polymerase II promoter
morphogenesis of a branching structure
maternal placenta development
heart valve development
ventricular septum development
transcription, DNA-templated
germ cell development
post-embryonic development
positive regulation of gene expression
negative regulation of B cell proliferation
negative regulation of lipopolysaccharide-mediated signaling pathway
methylation
regulation of natural killer cell differentiation
regulation of extrathymic T cell differentiation
aorta development
eye photoreceptor cell development
cell fate commitment
positive regulation of B cell differentiation
artery morphogenesis
regulation of NK T cell differentiation
intestinal epithelial cell development
trophoblast giant cell differentiation
coronary vasculature development
sebum secreting cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
methyltransferase activity
histone deacetylase binding
metal ion binding
promoter-specific chromatin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06009963.hg.1 | TC06000844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009963.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009963.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009960.hg.1 | TC06000844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009960.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000424894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000424894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000424894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000450060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000450060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000450060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000489365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000489365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000489365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161880.hg.1 | TC0600008972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369089.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369091.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000369096.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000424894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000424894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000424894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000450060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000450060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000450060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000481163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000489365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000489365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000489365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | ENST00000613999.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_001198.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008972.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161877.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161877.hg.1 | NM_182907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161877.hg.1 | TC0600008972.hg.1 | Clariom D TC |

PRDM1
Genomic Map