Assay Details
| Gene Symbol: NTRK1-DYNC2H1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NTRK1-DYNC2H1_N17D85.0 | - | 63 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NTRK1-DYNC2H1 fusion| Gene Symbol: NTRK1-DYNC2H1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NTRK1-DYNC2H1_N17D85.0 | - | 63 | 69 | ||
Set Membership: |
|