Assay Details
| Gene Symbol: PWWP2A-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PWWP2A-ROS1_P1R36.0 | - | 71 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PWWP2A-ROS1 fusion| Gene Symbol: PWWP2A-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PWWP2A-ROS1_P1R36.0 | - | 71 | 100 | ||