Assay Details
| Gene Symbol: CCL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002984.3 | NP_002975.1 | 2 - 3 | 269 | 61 | |
| GenBank mRNA | AY766446.1 | - | 2 - 3 | 269 | 61 | |
| BC104226.1 | - | 2 - 3 | 260 | 40032168 | 61 | |
| BC104227.1 | - | 2 - 3 | 260 | 40032172 | 61 | |
| BC107433.1 | - | 2 - 3 | 270 | 6727378 | 61 | |
| CR542119.1 | - | 2 - 3 | 190 | 61 | ||
| J04130.1 | - | 2 - 3 | 298 | 61 | ||
| M23502.1 | - | 2 - 3 | 264 | 61 | ||
| M25316.1 | - | 2 - 3 | 263 | 61 | ||
| X16166.1 | - | 2 - 3 | 242 | 61 | ||
| X53683.1 | - | 2 - 3 | 256 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CCL4
Genomic Map