Assay Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178329.2 | NP_847899.1 | 1 - 2 | 155 | 109 | |
| XM_011533335.2 | XP_011531637.1 | 2 - 3 | 341 | 109 | ||
| GenBank mRNA | AF224495.1 | - | 1 - 2 | 76 | 109 | |
| AF262299.1 | - | 1 - 2 | 105 | 109 | ||
| BC110297.1 | - | 1 - 2 | 64 | 5212541 | 109 | |
| BC130318.1 | - | 1 - 2 | 66 | 40146349 | 109 | |
| BC130320.1 | - | 1 - 2 | 66 | 40146351 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF224495.1, AF262299.1, BC110297.1, BC130318.1, BC130320.1, NM_178329.2, XM_011533335.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
chemotaxis
inflammatory response
cellular defense response
cell adhesion
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
viral process
positive regulation of angiogenesis
chemokine-mediated signaling pathway
chemokine receptor activity
protein binding
C-C chemokine receptor activity
positive regulation of endothelial cell proliferation
chemotaxis
inflammatory response
cellular defense response
cell adhesion
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
viral process
positive regulation of angiogenesis
chemokine-mediated signaling pathway
chemokine receptor activity
protein binding
C-C chemokine receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000357422.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000395942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000452454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000457243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000475150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000484025.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | ENST00000545097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001164680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_001837.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007255.hg.1 | NM_178329.2 | Refseq |

CCR3
Genomic Map