Assay Details
Gene Symbol: POU1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000306.3 | NP_000297.1 | 1 - 2 | 268 | 88 | |
GenBank mRNA | D10216.1 | - | 1 - 2 | 268 | 88 | |
L18781.1 | - | 1 - 2 | 143 | 88 | ||
X62429.1 | - | 1 - 2 | 162 | 88 | ||
X72215.1 | - | 1 - 2 | 143 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001585.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell fate specification
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
B cell differentiation
positive regulation of inositol trisphosphate biosynthetic process
positive regulation of multicellular organism growth
regulation of insulin-like growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nuclear transport
somatotropin secreting cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
negative regulation of transcription from RNA polymerase II promoter
cell fate specification
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
B cell differentiation
positive regulation of inositol trisphosphate biosynthetic process
positive regulation of multicellular organism growth
regulation of insulin-like growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nuclear transport
somatotropin secreting cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03028649.hg.1 | TC03001585.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03028649.hg.1 | ENST00000344265.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03028649.hg.1 | ENST00000350375.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03028649.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03028649.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03028649.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300187180.hg.1 | TC0300011712.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000344265.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000350375.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000560656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | ENST00000561167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_000306.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011712.hg.1 | NM_001122757.2 | Refseq |
