Assay Details
| Gene Symbol: MAGEB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277307.1 | NP_001264236.1 | 1 - 2 | 298 | 89 | |
| GenBank mRNA | DV080535.1 | - | 1 - 2 | 302 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277307.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | ENST00000400004.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006687.hg.1 | NM_001277307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000422438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000435789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009145.hg.1 | ENST00000454712.2 | Ensembl |

MAGEB17
Genomic Map