Assay Details
| Gene Symbol: SAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168682.1 | NP_001162153.1 | 4 - 5 | 399 | 72 | |
| XM_005250096.3 | XP_005250153.1 | 4 - 5 | 637 | 72 | ||
| XM_006715822.3 | XP_006715885.1 | 4 - 5 | 589 | 72 | ||
| GenBank mRNA | BQ635892.1 | - | 4 - 5 | 399 | 72 | |
More Information
Set Membership: |
|

SAP25
Genomic Map