Assay Details
| Gene Symbol: MTRNR2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190452.1 | NP_001177381.1 | 1 | 407 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190452.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NR_039705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000361390.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000386347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000387365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0M00006432.hg.1 | ENST00000389680.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000361390.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000386347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387347.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000387365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0M00006432.hg.1 | ENST00000389680.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NR_039705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | NM_001190452.1 | Refseq |

MTRNR2L1
Genomic Map