Assay Details
| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001732.3 | NP_001001732.2 | 4 - 5 | 523 | 102 | |
| NM_001159747.1 | NP_001153219.1 | 5 - 6 | 556 | 102 | ||
| XM_011539789.2 | XP_011538091.1 | 12 - 13 | 1617 | 102 | ||
| XM_011539790.2 | XP_011538092.1 | 12 - 13 | 1617 | 102 | ||
| XM_011539791.2 | XP_011538093.1 | 11 - 12 | 1434 | 102 | ||
| XM_011539792.2 | XP_011538094.1 | 6 - 7 | 739 | 102 | ||
| XM_011539793.2 | XP_011538095.1 | 5 - 6 | 1718 | 102 | ||
| XM_011539795.2 | XP_011538097.1 | 4 - 5 | 1535 | 102 | ||
| XM_017016240.1 | XP_016871729.1 | 5 - 6 | 795 | 102 | ||
| GenBank mRNA | AK302377.1 | - | 5 - 6 | 535 | 102 | |
| BC075861.1 | - | 4 - 5 | 488 | 5171414 | 102 | |
More Information
Set Membership: |
|

CC2D2B
Genomic Map