Assay Details
Gene Symbol: B3GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017007402.1 | XP_016862891.1 | 2 - 3 | 241 | 147 | |
XM_017007403.1 | XP_016862892.1 | 2 - 3 | 189 | 147 | ||
XM_017007404.1 | XP_016862893.1 | 2 - 3 | 197 | 147 | ||
XM_017007405.1 | XP_016862894.1 | 2 - 3 | 189 | 147 | ||
XM_017007411.1 | XP_016862900.1 | 2 - 3 | 337 | 147 | ||
GenBank mRNA | AK223439.1 | - | 2 - 3 | 136 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017007402.1, XM_017007403.1, XM_017007404.1, XM_017007405.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
