Assay Details
Gene Symbol: LOC100506393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040098.1 | 1 - 2 | 337 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | AE binding protein 2 | AEBP2 | -492.4kb | Sense | Chr. 12 - 19404101 - 19522239 on NCBI Build GRCh38 | |
Downstream | phosphodiesterase 3A | PDE3A |
CGI-PDE, CGI-PDE A, CGI-PDE-A, HTNB |
349.5kb | Sense | Chr. 12 - 20369245 - 20684107 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
