Assay Details
Gene Symbol: LOC105375734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131202.1 | 1 - 2 | 57 | 112 | ||
GenBank mRNA | DA599964.1 | - | 1 - 2 | 59 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HAS2 antisense RNA 1 | HAS2-AS1 |
HAS2-AS, HAS2AS, HASNT, NCRNA00077 |
-922.3kb | Anti-sense | Chr. 8 - 121639346 - 121645325 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1151 | LINC01151 | 101.7kb | Sense | Chr. 8 - 122670385 - 122694106 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
