Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
ENST00000598079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
ENST00000598079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
ENST00000598079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
ENST00000598079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
ENST00000598079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
ENST00000598079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008606.hg.1 |
NR_131205.1 |
Refseq |