Assay Details
Gene Symbol: XLOC_009911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131215.1 | 1 - 2 | 79 | 72 | ||
GenBank mRNA | HY014193.1 | - | 1 - 2 | 80 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | signal peptide peptidase like 3 | SPPL3 |
IMP2, MDHV1887, PRO4332, PSH1, PSL4 |
-343.0 | Anti-sense | Chr. 12 - 120762510 - 120904352 on NCBI Build GRCh38 |
Downstream | HNF1A antisense RNA 1 | HNF1A-AS1 |
C12orf27, NCRNA00262 |
61.8kb | Anti-sense | Chr. 12 - 120969838 - 120972292 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
