Assay Details
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131769.1 | 2 - 3 | 278 | 88 | ||
GenBank mRNA | AK131381.1 | - | 2 - 3 | 278 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fatty acid binding protein 6 | FABP6 |
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP |
-41.8kb | Anti-sense | Chr. 5 - 160187367 - 160238722 on NCBI Build GRCh38 |
Downstream | C1q and tumor necrosis factor related protein 2 | C1QTNF2 |
CTRP2, zacrp2 |
32.1kb | Sense | Chr. 5 - 160347558 - 160370656 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000257536.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000377503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000393977.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000519673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000520748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | ENST00000541762.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_001308173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NM_024565.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013392.hg.1 | NR_131769.1 | Refseq |
