Assay Details
| Gene Symbol: LINC00566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131903.1 | 1 - 2 | 94 | 100 | ||
| GenBank mRNA | BC038727.1 | - | 1 - 2 | 94 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C1q and tumor necrosis factor related protein 9 | C1QTNF9 |
AQL1, C1QTNF9A, CTRP9 |
-8.9kb | Sense | Chr. 13 - 24307166 - 24322531 on NCBI Build GRCh38 |
| Downstream | poly(ADP-ribose) polymerase family member 4 | PARP4 |
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193 |
83.8kb | Anti-sense | Chr. 13 - 24420930 - 24512993 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00566
Genomic Map