Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
NR_132371.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
NR_132371.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007664.hg.1 |
NR_132371.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000457672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
ENST00000609962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
NR_132371.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
NR_132371.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007664.hg.1 |
NR_132371.1 |
Refseq |