Assay Details
Gene Symbol: LOC105376575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132751.1 | 1 - 2 | 2440 | 90 | ||
GenBank mRNA | AK096475.1 | - | 1 - 2 | 2440 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleobindin 2 | NUCB2 |
HEL-S-109, NEFA |
-161.0 | Sense | Chr. 11 - 17260340 - 17331881 on NCBI Build GRCh38 |
Downstream | natural killer cell cytotoxicity receptor 3 ligand 1 | NCR3LG1 |
B7-H6, B7H6, DKFZp686O24166 |
14.2kb | Sense | Chr. 11 - 17351762 - 17377321 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
