Assay Details
| Gene Symbol: SNORA90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132772.1 | 1 | 112 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | apoptosis antagonizing transcription factor | AATF |
BFR2, CHE-1, CHE1, DED |
-187.9kb | Sense | Chr. 17 - 36948875 - 37056871 on NCBI Build GRCh38 |
| Downstream | chromosome 17 open reading frame 78 | C17orf78 | 130.7kb | Sense | Chr. 17 - 37375710 - 37392708 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORA90
Genomic Map