Assay Details
| Gene Symbol: S100A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_133645.1 | 2 - 3 | 399 | 144 | ||
| GenBank mRNA | AW954845.1 | - | 2 - 3 | 375 | 144 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | S100 calcium binding protein A2 | S100A2 |
CAN19, S100L |
-41.1kb | Sense | Chr. 1 - 153561108 - 153565830 on NCBI Build GRCh38 |
| Downstream | S100 calcium binding protein A14 | S100A14 |
BCMP84, S100A15 |
6.1kb | Sense | Chr. 1 - 153614255 - 153616917 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368705.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000368706.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | ENST00000474991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015871.hg.1 | NM_080388.2 | Refseq |

S100A16
Genomic Map