Assay Details
Gene Symbol: DDX39B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_133675.1 | 1 - 2 | 93 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP6V1G2-DDX39B readthrough (NMD candidate) | ATP6V1G2-DDX39B | Anti-sense | Chr. 6 - 31530219 - 31546848 on NCBI Build GRCh38 | ||
Downstream | ATPase H+ transporting V1 subunit G2 | ATP6V1G2 |
ATP6G, ATP6G2, NG38, VMA10 |
1.3kb | Anti-sense | Chr. 6 - 31544451 - 31546848 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
