Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008743.hg.1 |
ENST00000513958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008743.hg.1 |
ENST00000513958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008743.hg.1 |
ENST00000513958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010564.hg.1 |
ENST00000565901.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008743.hg.1 |
ENST00000513958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008743.hg.1 |
ENST00000513958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008743.hg.1 |
ENST00000513958.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010564.hg.1 |
ENST00000565901.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010564.hg.1 |
ENST00000567058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000548359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000551032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008285.hg.1 |
ENST00000552167.1 |
Ensembl |