Assay Details
| Gene Symbol: TRPC7-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_133682.1 | 3 - 4 | 381 | 164 | ||
| GenBank mRNA | BI765200.1 | - | 3 - 4 | 381 | 164 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC389332 | LOC389332 | -121.0kb | Anti-sense | Chr. 5 - 136191468 - 136193163 on NCBI Build GRCh38 | |
| Downstream |
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1 |
SPOCK1 |
SPOCK, TESTICAN, TIC1 |
659.2kb | Anti-sense | Chr. 5 - 136975298 - 137499329 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

TRPC7-AS2
Genomic Map