Assay Details
| Gene Symbol: LOC101927278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134451.1 | 3 - 4 | 673 | 99 | ||
| NR_134452.1 | 3 - 4 | 669 | 99 | |||
| NR_134453.1 | 2 - 3 | 638 | 99 | |||
| NR_134454.1 | 2 - 3 | 634 | 99 | |||
| GenBank mRNA | BE501771.1 | - | 1 - 2 | 99 | ||
| BX103104.1 | - | 2 - 3 | 593 | 99 | ||
| CB123723.1 | - | 3 - 4 | 348 | 99 | ||
| EL948012.1 | - | 2 - 3 | 259 | 99 | ||
| HY194333.1 | - | 1 - 2 | 99 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HPS1, biogenesis of lysosomal organelles complex 3 subunit 1 |
HPS1 |
BLOC3S1, HPS |
-6.2kb | Anti-sense | Chr. 10 - 98415068 - 98446963 on NCBI Build GRCh38 |
| Downstream | heparanase 2 (inactive) | HPSE2 |
HPA2, HPR2, UFS, UFS1 |
3.3kb | Anti-sense | Chr. 10 - 98457077 - 99235875 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927278
Genomic Map