Assay Details
| Gene Symbol: LOC105378269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134498.1 | 1 - 2 | 284 | 85 | ||
| GenBank mRNA | BM682592.1 | - | 1 - 2 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1518 | LINC01518 | -63.4kb | Anti-sense | Chr. 10 - 42673013 - 42691759 on NCBI Build GRCh38 | |
| Downstream |
BMS1, ribosome biogenesis factor |
BMS1 |
ACC, BMS1L |
12.9kb | Sense | Chr. 10 - 42782506 - 42834937 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378269
Genomic Map