Assay Details
Gene Symbol: LOC105378123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134592.1 | 1 - 2 | 54 | 81 | ||
GenBank mRNA | BX109291.1 | - | 1 - 2 | 54 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | G protein-coupled receptor 31 | GPR31 |
12-HETER, HETER, HETER1 |
-8.4kb | Sense | Chr. 6 - 167156872 - 167157831 on NCBI Build GRCh38 |
Downstream | t-complex 10-like 2 | TCP10L2 | bA517H2.3 | 613.0 | Anti-sense | Chr. 6 - 167170593 - 167196777 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
