Assay Details
Gene Symbol: LOC105374960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134615.1 | 1 - 2 | 95 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928519 | LOC101928519 | -269.3kb | Anti-sense | Chr. 6 - 19068543 - 19180480 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC100506885 | LOC100506885 | 270.1kb | Anti-sense | Chr. 6 - 19729421 - 19804759 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
