Assay Details
Gene Symbol: LOC105373006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134617.1 | 2 - 3 | 223 | 87 | ||
GenBank mRNA | DB089380.1 | - | 2 - 3 | 223 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | synapsin III | SYN3 | -50.7kb | Anti-sense | Chr. 22 - 32508022 - 33058391 on NCBI Build GRCh38 | |
Downstream | LARGE xylosyl- and glucuronyltransferase 1 | LARGE1 |
LARGE, MDC1D, MDDGA6, MDDGB6 |
51.6kb | Anti-sense | Chr. 22 - 33162237 - 33922841 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
